Assay Details
Target Gene Details
Entrez Gene ID: | 91612 |
Gene Name: | churchill domain containing 1 |
Gene Aliases: |
C14orf52, My015, chch |
Location: |
Chr.14:64914361-64935368 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHURC1 | NM_001204063.1 | 4 | 1932 | NP_001190992.1 |
| NM_001204064.1 | 3 | 1861 | NP_001190993.1 | |
| NM_145165.3 | 4 | 1935 | NP_660148.3 | |
| AK057626.1 | 4 | 1843 | ||
| AK296141.1 | ||||
| AK315905.1 | ||||
| BC020550.1 | 4 | 1881 | AAH20550.1 |
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
| NM_001202559.1 | NP_001189488.1 | |||
| AK296850.1 | ||||
| AK303739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map