Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL1 | NM_005576.3 | 1 | 198 | NP_005567.2 |
| XM_011521555.1 | 1 | 205 | XP_011519857.1 | |
| AK303982.1 | 1 | 194 | ||
| AK314222.1 | 1 | 196 | ||
| AL529650.3 | 1 | 224 | ||
| BC015090.2 | 1 | 177 | AAH15090.1 | |
| BC068542.1 | 1 | 198 | AAH68542.1 | |
| L21186.1 | 1 | 177 | AAA50162.1 |
Target Gene Details
Entrez Gene ID: | 100287616 |
Gene Name: | LOXL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:73917468-73928248 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL1-AS1 | NR_040066.1 | |||
| NR_040067.1 | ||||
| NR_040068.1 | ||||
| NR_040069.1 | ||||
| NR_040070.1 | ||||
| AK056376.1 | ||||
| AK056885.1 | ||||
| BX355383.2 | ||||
| BX366015.2 | ||||
| DA021396.1 | ||||
| DA038997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion |
|
| nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion |
|
| nsv428308 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map