Assay Details
Target Gene Details
Entrez Gene ID: | 1271 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
- |
Location: |
Chr.9:34551432-34590386 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTFR | NM_001207011.1 | NP_001193940.1 | ||
| NM_001842.4 | NP_001833.1 | |||
| NM_147164.2 | NP_671693.1 | |||
| XM_017014260.1 | XP_016869749.1 | |||
| XM_017014261.1 | XP_016869750.1 | |||
| XM_017014262.1 | XP_016869751.1 | |||
| XM_017014263.1 | XP_016869752.1 | |||
| XM_017014264.1 | XP_016869753.1 | |||
| XM_017014265.1 | XP_016869754.1 | |||
| AK127444.1 | ||||
| BC001492.2 | AAH01492.1 | |||
| BT019824.1 | AAV38627.1 | |||
| BX364434.2 | ||||
| HQ628632.1 | ||||
| M73238.1 | AAA35707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion |
|
| nsv831543 | Chr.9:34431291 - 34615384 on Build GRCh38 | Loss |
|
| nsv519946 | Chr.9:34521869 - 34555001 on Build GRCh38 | Loss |
|
| nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map