Assay Details
Target Gene Details
Entrez Gene ID: | 10614 |
Gene Name: | hexamethylene bisacetamide inducible 1 |
Gene Aliases: |
CLP1, EDG1, HIS1, MAQ1 |
Location: |
Chr.17:45147317-45152101 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXIM1 | NM_006460.2 | 1 | 2017 | NP_006451.1 |
| AB021179.1 | 1 | 832 | BAA36166.1 | |
| AK023624.1 | 1 | 859 | ||
| AK313557.1 | 1 | 374 | ||
| BC006460.1 | 1 | 750 | AAH06460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2062 | Chr.17:45145375 - 45172616 on Build GRCh38 | Insertion |
|
| nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss |
|
| nsv828022 | Chr.17:45137483 - 45160552 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map