Assay Details
Target Gene Details
Entrez Gene ID: | 2980 |
Gene Name: | guanylate cyclase activator 2A |
Gene Aliases: |
GCAP-I, GUCA2, STARA |
Location: |
Chr.1:42162691-42164724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCA2A | NM_033553.2 | NP_291031.2 | ||
| M95174.1 | AAA58625.1 | |||
| M97496.1 | AAA35915.1 | |||
| T28160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527531 | Chr.1:42151361 - 42169834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map