Assay Details
Target Gene Details
Entrez Gene ID: | 2934 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF, AGEL |
Location: |
Chr.9:121201082-121332844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSN | XM_011518591.2 | XP_011516893.1 | ||
| XM_017014643.1 | XP_016870132.1 | |||
| XM_017014644.1 | XP_016870133.1 | |||
| XM_017014647.1 | XP_016870136.1 | |||
| AK125819.1 |
Target Gene Details
Entrez Gene ID: | 51552 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
FBP, RAB-14 |
Location: |
Chr.9:121178137-121202087 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB14 | NM_016322.3 | 1 | 247 | NP_057406.2 |
| AK023524.1 | BAB14598.1 | |||
| AK090889.1 | 1 | 46 | ||
| AL162081.1 | CAB82414.1 | |||
| BC006081.1 | AAH06081.1 | |||
| DA404780.1 | 1 | 247 |
More Information
Set Membership: |
|

Genomic Map