Assay Details
Target Gene Details
Entrez Gene ID: | 11180 |
Gene Name: | WD repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.3:49007062-49015953 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR6 | NM_001320546.1 | NP_001307475.1 | ||
| NM_018031.4 | NP_060501.3 | |||
| AF099100.1 | AAF80244.1 | |||
| AK001080.1 | BAA91497.1 | |||
| AK092079.1 | ||||
| AK225653.1 | ||||
| AK295145.1 | ||||
| AK296387.1 | ||||
| AK297090.1 | ||||
| AK309913.1 | ||||
| BC002826.2 | AAH02826.1 | |||
| BC101707.1 | AAI01708.1 | |||
| BC113467.1 | ||||
| DC377068.1 | ||||
| DC389885.1 | ||||
| DC400613.1 | ||||
| DC408104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map