Assay Details
Target Gene Details
Entrez Gene ID: | 1828 |
Gene Name: | desmoglein 1 |
Gene Aliases: |
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1 |
Location: |
Chr.18:31318089-31359246 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1 | NM_001942.3 | 12 | 1981 | NP_001933.2 |
| AF097935.1 | 12 | 1982 | AAC83817.1 | |
| X56654.1 | 12 | 1846 | CAA39976.1 |
Target Gene Details
Entrez Gene ID: | 101927718 |
Gene Name: | DSG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:31343368-31426988 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1-AS1 | NR_110788.1 | 4 | 567 | |
| NR_110789.1 | 3 | 306 | ||
| BX476374.1 | 4 | 507 | ||
| DB526502.1 | 3 | 306 |
More Information
Set Membership: |
|

Genomic Map