Assay Details
Target Gene Details
Entrez Gene ID: | 84181 |
Gene Name: | chromodomain helicase DNA binding protein 6 |
Gene Aliases: |
CHD-6, CHD5, RIGB |
Location: |
Chr.20:41402101-41618494 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD6 | NM_032221.4 | 37 | 7820 | NP_115597.3 |
| XM_005260573.2 | 36 | 7642 | XP_005260630.1 | |
| XM_005260576.4 | 14 | 3954 | XP_005260633.1 | |
| XM_011529080.2 | 36 | 7881 | XP_011527382.1 | |
| XM_011529082.2 | 26 | 6053 | XP_011527384.1 | |
| XM_017028099.1 | 36 | 9316 | XP_016883588.1 | |
| XM_017028100.1 | 36 | 7841 | XP_016883589.1 | |
| XM_017028101.1 | 35 | 9172 | XP_016883590.1 | |
| XM_017028103.1 | 34 | 7275 | XP_016883592.1 | |
| AB037756.2 | 23 | 5621 | BAA92573.2 | |
| AK098144.1 | 3 | 2253 | ||
| AY034072.1 | 37 | 7820 | AAK56405.1 | |
| BC021907.1 | 1 | 321 | AAH21907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss |
|
| nsv3381 | Chr.20:41296686 - 42137056 on Build GRCh38 | Deletion |
|
| nsv478389 | Chr.20:41405075 - 41405075 on Build GRCh38 | Novel Sequence Insertion |
|
| nsv833984 | Chr.20:41239403 - 41417792 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map