Assay Details
Target Gene Details
Entrez Gene ID: | 91543 |
Gene Name: | radical S-adenosyl methionine domain containing 2 |
Gene Aliases: |
2510004L01Rik, cig33, cig5, vig1 |
Location: |
Chr.2:6865733-6898239 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSAD2 | NM_080657.4 | 3 | 758 | NP_542388.2 |
| XM_011510414.2 | 4 | 1847 | XP_011508716.1 | |
| XM_011510415.2 | 3 | 589 | XP_011508717.1 | |
| AF442151.1 | 3 | 646 | AAL50053.1 | |
| AK300085.1 | 3 | 524 | ||
| AK312670.1 | 3 | 657 | ||
| BC017969.1 | 3 | 646 | AAH17969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map