Assay Details
Target Gene Details
Entrez Gene ID: | 57696 |
Gene Name: | DEAD-box helicase 55 |
Gene Aliases: |
- |
Location: |
Chr.12:123602077-123620943 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX55 | NM_020936.2 | 9 | 948 | NP_065987.1 |
| NR_135104.1 | 9 | 948 | ||
| NR_135105.1 | 9 | 948 | ||
| XM_011538608.2 | 9 | 990 | XP_011536910.1 | |
| XM_017019710.1 | 9 | 987 | XP_016875199.1 | |
| XM_017019711.1 | 8 | 1042 | XP_016875200.1 | |
| XM_017019712.1 | 9 | 840 | XP_016875201.1 | |
| XM_017019713.1 | 9 | 963 | XP_016875202.1 | |
| XM_017019714.1 | 10 | 1121 | XP_016875203.1 | |
| XM_017019715.1 | 9 | 988 | XP_016875204.1 | |
| XM_017019716.1 | 9 | 933 | XP_016875205.1 | |
| XM_017019717.1 | 9 | 885 | XP_016875206.1 | |
| XM_017019718.1 | 7 | 1303 | XP_016875207.1 | |
| XM_017019719.1 | 9 | 840 | XP_016875208.1 | |
| XM_017019720.1 | 7 | 1918 | XP_016875209.1 | |
| XM_017019721.1 | 9 | 934 | XP_016875210.1 | |
| AB046815.1 | 4 | 2775 | BAB13421.1 | |
| AK074219.1 | 9 | 900 | BAB85021.1 | |
| AK131442.1 | 6 | 1187 | ||
| AK301043.1 | 9 | 911 | ||
| AK307384.1 | 9 | 924 | ||
| AL833756.1 | 7 | 1985 | CAH56233.1 | |
| BC030020.2 | 9 | 892 | AAH30020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain |
|
| dgv524e212 | Chr.12:123614065 - 123625575 on Build GRCh38 | Loss |
|
| nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map