Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map