Assay Details
Target Gene Details
Entrez Gene ID: | 5155 |
Gene Name: | platelet derived growth factor subunit B |
Gene Aliases: |
IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis |
Location: |
Chr.22:39223359-39245055 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFB | NM_002608.3 | 7 | 2233 | NP_002599.1 |
| NM_033016.3 | 7 | 1133 | NP_148937.1 | |
| AK022920.1 | 1 | 337 | ||
| BC029822.1 | 7 | 1453 | AAH29822.1 | |
| M12783.1 | 7 | 2233 | AAA60553.1 | |
| M32009.1 | 1 | 136 | ||
| X02744.1 | 7 | 1258 | CAA26524.1 | |
| X02811.1 | CAA26579.1 | |||
| X03702.1 | 6 | 1066 | CAA27333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647761 | Chr.22:39204243 - 39286742 on Build GRCh38 | Gain |
|
| nsv1160762 | Chr.22:39214849 - 39298220 on Build GRCh38 | Deletion |
|
| nsv834198 | Chr.22:39222748 - 39409818 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116011535] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map