Assay Details
Target Gene Details
Entrez Gene ID: | 401 |
Gene Name: | paired like homeobox 2a |
Gene Aliases: |
ARIX, CFEOM2, FEOM2, NCAM2, PMX2A |
Location: |
Chr.11:72239077-72244176 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHOX2A | NM_005169.3 | 3 | 1195 | NP_005160.2 |
| AK290645.1 | 3 | 1195 | ||
| BC041564.1 | 3 | 1177 | AAH41564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951038 | Chr.11:72226157 - 72253456 on Build GRCh38 | Deletion |
|
| nsv555405 | Chr.11:72233623 - 72241301 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map