Assay Details
Target Gene Details
Entrez Gene ID: | 6564 |
Gene Name: | solute carrier family 15 member 1 |
Gene Aliases: |
HPECT1, HPEPT1, PEPT1 |
Location: |
Chr.13:98683801-98752675 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC15A1 | NM_005073.3 | 23 | 2211 | NP_005064.1 |
| AF043233.1 | 23 | 2190 | AAC15477.1 | |
| AY634368.1 | 23 | 2181 | AAT48121.1 | |
| BC096327.1 | 23 | 2165 | AAH96327.1 | |
| BC096328.3 | 23 | 2165 | AAH96328.1 | |
| BC096329.3 | 23 | 2165 | AAH96329.1 | |
| CU445953.1 | 1 | 582 | ||
| EU544224.1 | ||||
| U13173.1 | 23 | 2190 | AAB61693.1 | |
| U21936.1 | 23 | 2190 | AAA63797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038016 | Chr.13:98665177 - 98817733 on Build GRCh38 | Gain |
|
| esv2747861 | Chr.13:98625149 - 98716478 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map