Assay Details
Target Gene Details
Entrez Gene ID: | 28511 |
Gene Name: | NFKB inhibitor interacting Ras like 2 |
Gene Aliases: |
KBRAS2, kappaB-Ras2 |
Location: |
Chr.17:42017576-42025641 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKIRAS2 | NM_001001349.2 | 4 | 2178 | NP_001001349.1 |
| NM_001144927.1 | 4 | 2282 | NP_001138399.1 | |
| NM_001144928.1 | 4 | 2095 | NP_001138400.1 | |
| NM_001144929.1 | 5 | 2166 | NP_001138401.1 | |
| NM_017595.5 | 4 | 2263 | NP_060065.2 | |
| XM_005257251.1 | 4 | 2290 | XP_005257308.1 | |
| AL137682.1 | 2 | 4003 | CAB70873.2 | |
| BC063498.1 | 4 | 2734 | AAH63498.1 |
More Information
Set Membership: |
|

Genomic Map