Assay Details
Target Gene Details
Entrez Gene ID: | 30835 |
Gene Name: | CD209 molecule |
Gene Aliases: |
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1 |
Location: |
Chr.19:7739993-7747609 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD209 | NM_001144893.1 | 5 | 1840 | NP_001138365.1 |
| NM_001144894.1 | 5 | 2116 | NP_001138366.1 | |
| NM_001144895.1 | 7 | 1972 | NP_001138367.1 | |
| NM_001144896.1 | 6 | 2176 | NP_001138368.1 | |
| NM_001144897.1 | 7 | 2230 | NP_001138369.1 | |
| NM_001144899.1 | 7 | 1765 | NP_001138371.1 | |
| NM_021155.3 | 7 | 2248 | NP_066978.1 | |
| NR_026692.1 | 6 | 2348 | ||
| XM_005272472.4 | 7 | 1865 | XP_005272529.1 | |
| AF290886.1 | 7 | 2190 | AAK20997.1 | |
| AK057212.1 | 1 | 390 | ||
| AK293089.1 | 7 | 2204 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
|
| nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain |
|
| esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
|
| nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map