Assay Details
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUMA1 | NM_001286561.1 | 27 | 7215 | NP_001273490.1 |
| NM_006185.3 | 27 | 7159 | NP_006176.2 | |
| NR_104476.1 | 25 | 3670 | ||
| XM_006718564.1 | 26 | 6974 | XP_006718627.1 | |
| XM_011545054.1 | 27 | 7028 | XP_011543356.1 | |
| XM_011545055.1 | 28 | 7206 | XP_011543357.1 | |
| XM_011545056.2 | 29 | 7200 | XP_011543358.1 | |
| XM_011545057.2 | 29 | 7190 | XP_011543359.1 | |
| XM_011545058.1 | 27 | 7227 | XP_011543360.1 | |
| XM_011545059.2 | 29 | 7192 | XP_011543361.1 | |
| XM_011545060.2 | 29 | 7186 | XP_011543362.1 | |
| XM_011545061.2 | 27 | 7046 | XP_011543363.1 | |
| XM_011545062.2 | 27 | 7059 | XP_011543364.1 | |
| XM_011545063.2 | 27 | 7026 | XP_011543365.1 | |
| XM_011545064.1 | 27 | 7013 | XP_011543366.1 | |
| XM_011545065.1 | 26 | 6986 | XP_011543367.1 | |
| XM_011545066.1 | 25 | 6932 | XP_011543368.1 | |
| AB208841.1 | 12 | 5232 | BAD92078.1 | |
| AB210007.1 | 27 | 6960 | ||
| BC004165.2 | 25 | 3585 | AAH04165.1 | |
| BC008345.1 | 13 | 3367 | AAH08345.1 | |
| BC013023.2 | 12 | 4061 | AAH13023.1 | |
| BG754891.1 | 1 | 485 | ||
| Z11583.1 | 27 | 6977 | CAA77669.1 | |
| Z11584.1 | 27 | 7037 | CAA77670.1 | |
| Z14227.1 | 8 | 4558 | ||
| Z14228.1 | 7 | 2630 | ||
| Z14229.1 | 10 | 2096 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
| nsv951037 | Chr.11:72000055 - 72016354 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map