Assay Details
Target Gene Details
Entrez Gene ID: | 23265 |
Gene Name: | exocyst complex component 7 |
Gene Aliases: |
2-5-3p, EX070, EXO70, EXOC1, Exo70p, YJL085W |
Location: |
Chr.17:76080993-76104221 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOC7 | NM_001013839.3 | 19 | 4029 | NP_001013861.1 |
| NM_001145297.3 | 20 | 4182 | NP_001138769.1 | |
| NM_001145298.3 | 19 | 4005 | NP_001138770.1 | |
| NM_001145299.3 | 20 | 4098 | NP_001138771.1 | |
| NM_001282313.1 | 19 | 4058 | NP_001269242.1 | |
| NM_015219.4 | 18 | 3936 | NP_056034.2 | |
| XM_006721786.3 | 21 | 4136 | XP_006721849.1 | |
| XM_006721787.3 | 20 | 4502 | XP_006721850.1 | |
| XM_006721788.3 | 20 | 4043 | XP_006721851.1 | |
| XM_006721789.3 | 19 | 3974 | XP_006721852.1 | |
| XM_011524571.2 | 20 | 4413 | XP_011522873.1 | |
| XM_017024397.1 | 16 | 3538 | XP_016879886.1 | |
| AB028990.1 | 19 | 3952 | ||
| AK000718.1 | 1 | 119 | ||
| AK022552.1 | 10 | 2720 | BAB14095.1 | |
| AK023832.1 | 18 | 3844 | BAB14694.1 | |
| AK024769.1 | 1 | 1894 | ||
| AK128278.1 | 6 | 2176 | ||
| AL162005.1 | 1 | 1471 | ||
| BC015165.1 | 1 | 377 | AAH15165.1 | |
| BX648435.1 | 3 | 2585 |
Target Gene Details
Entrez Gene ID: | 353174 |
Gene Name: | zinc activated ion channel |
Gene Aliases: |
L2, LGICZ, LGICZ1, ZAC, ZAC1 |
Location: |
Chr.17:76079182-76082804 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZACN | NM_180990.3 | NP_851321.2 | ||
| AB223030.1 | BAE19924.1 | |||
| AF512521.1 | AAO20969.1 | |||
| AK122638.1 | ||||
| BC110597.1 | AAI10598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2142 | Chr.17:76070185 - 76087802 on Build GRCh38 | Insertion |
|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
| nsv509674 | Chr.17:76052197 - 76106436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map