Assay Details
Target Gene Details
Entrez Gene ID: | 23315 |
Gene Name: | solute carrier family 9 member A8 |
Gene Aliases: |
NHE-8, NHE8 |
Location: |
Chr.20:49812713-49892242 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646017 | Chr.20:49610386 - 49857106 on Build GRCh38 | Gain |
|
| esv3646034 | Chr.20:49788665 - 49826482 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73910322] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map