Assay Details
Target Gene Details
Entrez Gene ID: | 101929664 |
Gene Name: | HORMAD2 antisense RNA 1 |
Gene Aliases: |
MTMR3-AS1 |
Location: |
Chr.22:30008742-30080480 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HORMAD2-AS1 | NR_110541.1 | |||
| BG618381.1 |
Target Gene Details
Entrez Gene ID: | 8897 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
FYVE-DSP1, ZFYVE10 |
Location: |
Chr.22:29883165-30030868 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR3 | NM_021090.3 | 20 | 3876 | NP_066576.1 |
| NM_153050.2 | 20 | 3792 | NP_694690.1 | |
| NM_153051.2 | 19 | 3765 | NP_694691.1 | |
| XM_005261803.2 | 22 | 4021 | XP_005261860.1 | |
| XM_005261804.3 | 21 | 3938 | XP_005261861.1 | |
| XM_005261805.2 | 21 | 3994 | XP_005261862.1 | |
| XM_005261806.2 | 21 | 3903 | XP_005261863.1 | |
| XM_005261808.2 | 20 | 3883 | XP_005261865.1 | |
| XM_017029028.1 | 20 | 3795 | XP_016884517.1 | |
| XM_017029029.1 | 19 | 3758 | XP_016884518.1 | |
| XM_017029030.1 | 19 | 3690 | XP_016884519.1 | |
| XM_017029031.1 | 18 | 3651 | XP_016884520.1 | |
| XM_017029032.1 | 19 | 3648 | XP_016884521.1 | |
| XM_017029033.1 | 15 | 3222 | XP_016884522.1 | |
| XM_017029034.1 | 21 | 5225 | XP_016884523.1 | |
| AB002369.1 | 20 | 3795 | ||
| AF233436.2 | 17 | 3473 | AAF40203.2 | |
| AF233437.1 | 16 | 3446 | AAF40204.1 | |
| AF233438.1 | 17 | 3557 | AAF40205.1 | |
| AK289581.1 | 20 | 3876 | ||
| BC142713.1 | 19 | 3721 | ||
| BC148216.1 | 18 | 3584 | ||
| BC151217.1 | 20 | 3890 | ||
| BC152455.1 | 20 | 3795 | ||
| CR456525.1 | 18 | 3572 | CAG30411.1 |
More Information
Set Membership: |
|

Genomic Map