Assay Details
Target Gene Details
Entrez Gene ID: | 9481 |
Gene Name: | solute carrier family 25 member 27 |
Gene Aliases: |
UCP4 |
Location: |
Chr.6:46652915-46678190 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A27 | NM_001204051.1 | NP_001190980.1 | ||
| NM_001204052.1 | NP_001190981.1 | |||
| NM_004277.4 | NP_004268.3 | |||
| XM_005249483.2 | XP_005249540.1 | |||
| AK291427.1 | ||||
| AK295233.1 | ||||
| AK302906.1 | ||||
| BC033091.1 | AAH33091.1 | |||
| BP310559.1 | ||||
| DA144842.1 | ||||
| DC314392.1 |
More Information
Set Membership: |
|

Genomic Map