Assay Details
Target Gene Details
Entrez Gene ID: | 5709 |
Gene Name: | proteasome 26S subunit, non-ATPase 3 |
Gene Aliases: |
P58, RPN3, S3, TSTA2 |
Location: |
Chr.17:39980768-39997960 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD3 | NM_002809.3 | 2 | 507 | NP_002800.2 |
| AK022896.1 | 2 | 418 | ||
| AK054611.1 | 2 | 451 | ||
| AK055799.1 | 3 | 388 | BAB71019.1 | |
| AK094206.1 | 3 | 429 | BAC04310.1 | |
| AK293539.1 | 3 | 393 | ||
| AK298411.1 | ||||
| AK300081.1 | ||||
| AK300973.1 | 3 | 423 | ||
| BC000074.2 | 2 | 407 | AAH00074.1 | |
| BC004859.2 | 2 | 369 | AAH04859.1 | |
| BC012302.1 | 2 | 455 | AAH12302.1 | |
| BC020518.1 | 2 | 405 | AAH20518.1 | |
| BC025686.1 | 2 | 405 | AAH25686.1 | |
| BT007217.1 | 2 | 304 | AAP35881.1 | |
| CR456772.1 | 2 | 304 | CAG33053.1 | |
| D67025.1 | 2 | 461 | BAA23651.1 | |
| HY034626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map