Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG2 | NM_001282211.1 | NP_001269140.1 | ||
| AK293514.1 |
Target Gene Details
Entrez Gene ID: | 122664 |
Gene Name: | tubulin polymerization promoting protein family member 2 |
Gene Aliases: |
C14orf8, CT152, P18, p25beta |
Location: |
Chr.14:21024262-21036352 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPPP2 | NM_173846.4 | 2 | 230 | NP_776245.2 |
| XM_005267324.4 | 2 | 291 | XP_005267381.1 | |
| XM_011536416.1 | 2 | 166 | XP_011534718.1 | |
| XM_011536417.2 | 2 | 166 | XP_011534719.1 | |
| XM_011536418.1 | 2 | 166 | XP_011534720.1 | |
| XM_011536420.2 | 2 | 166 | XP_011534722.1 | |
| XM_017020966.1 | 2 | 194 | XP_016876455.1 | |
| AK093070.1 | 2 | 162 | ||
| AK098680.1 | 2 | 159 | ||
| AY072034.1 | 2 | 167 | AAL62338.1 | |
| AY173946.1 | 2 | 167 | AAO49716.1 | |
| BC038970.2 | 1 | 119 | AAH38970.1 | |
| DB458250.1 | 2 | 230 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map