Assay Details
Target Gene Details
Entrez Gene ID: | 56172 |
Gene Name: | ANKH inorganic pyrophosphate transport regulator |
Gene Aliases: |
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK |
Location: |
Chr.5:14704800-14871778 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKH | NM_054027.4 | NP_473368.1 | ||
| XM_017009644.1 | XP_016865133.1 | |||
| AB046801.1 | ||||
| AF274753.1 | AAF88039.1 | |||
| AK001799.1 | ||||
| AK130676.1 | ||||
| AK315012.1 | ||||
| AY358503.1 | AAQ88867.1 | |||
| BC009835.2 | AAH09835.1 | |||
| BC014526.2 | AAH14526.1 |
Target Gene Details
Entrez Gene ID: | 100130744 |
Gene Name: | uncharacterized LOC100130744 |
Gene Aliases: |
- |
Location: |
Chr.5:14712694-14716529 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130744 | NR_046285.1 | 1 | 985 | |
| AF289590.1 | 1 | 1004 | AAL55774.1 |
Target Gene Details
Entrez Gene ID: | 90268 |
Gene Name: | OTU deubiquitinase with linear linkage specificity |
Gene Aliases: |
FAM105B, GUM |
Location: |
Chr.5:14660794-14716552 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTULIN | XM_011514151.2 | 8 | 6094 | XP_011512453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026914 | Chr.5:14237673 - 14751479 on Build GRCh38 | Gain |
|
| nsv1018187 | Chr.5:14385055 - 15126994 on Build GRCh38 | Gain |
|
| nsv4728 | Chr.5:14691303 - 14725902 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map