Assay Details
Target Gene Details
Entrez Gene ID: | 1513 |
Gene Name: | cathepsin K |
Gene Aliases: |
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD |
Location: |
Chr.1:150796208-150808441 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSK | NM_000396.3 | 8 | 1271 | NP_000387.1 |
| AK130134.1 | 8 | 1090 | ||
| AK308229.1 | 7 | 932 | ||
| BC016058.1 | 8 | 1153 | AAH16058.1 | |
| S79895.1 | 8 | 1183 | AAB35521.1 | |
| U13665.1 | 8 | 1147 | AAA65233.1 | |
| U20280.1 | 8 | 1060 | AAA95998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2799 | Chr.1:150791723 - 150805959 on Build GRCh38 | Insertion |
|
| nsv437169 | Chr.1:150765721 - 150844410 on Build GRCh38 | Loss |
|
| nsv2788 | Chr.1:150782858 - 150828309 on Build GRCh38 | Deletion |
|
| nsv160180 | Chr.1:150792800 - 150800194 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map