Assay Details
Target Gene Details
Entrez Gene ID: | 4729 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit V2 |
Gene Aliases: |
CI-24k |
Location: |
Chr.18:9102623-9134345 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV2 | NM_021074.4 | 1 | 119 | NP_066552.2 |
| AK291865.1 | ||||
| AK312129.1 | ||||
| BC001632.1 | AAH01632.1 | |||
| BC017487.1 | AAH17487.1 | |||
| BC071689.1 | AAH71689.1 | |||
| BG742948.1 | 1 | 119 | ||
| CB121843.1 | 1 | 72 | ||
| CR456928.1 | CAG33209.1 | |||
| M22538.1 | AAA75390.1 |
More Information
Set Membership: |
|

Genomic Map