Assay Details
Target Gene Details
Entrez Gene ID: | 55304 |
Gene Name: | serine palmitoyltransferase long chain base subunit 3 |
Gene Aliases: |
C20orf38, LCB2B, LCB3, SPT3, SPTLC2L, dJ718P11, dJ718P11.1, hLCB2b |
Location: |
Chr.20:13008979-13169103 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTLC3 | NM_018327.2 | 1 | 222 | NP_060797.2 |
| XM_011529279.1 | XP_011527581.1 | |||
| XM_017027922.1 | XP_016883411.1 | |||
| AK001974.1 | BAA92012.1 | |||
| AK300184.1 | ||||
| BC005205.1 | 1 | 222 | ||
| BC150644.1 | 1 | 181 | ||
| CF994231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4247n100 | Chr.20:12748832 - 13357690 on Build GRCh38 | Gain |
|
| nsv518184 | Chr.20:12763161 - 13364025 on Build GRCh38 | Gain |
|
| nsv833923 | Chr.20:12924041 - 13083778 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map