Assay Details
Target Gene Details
Entrez Gene ID: | 5424 |
Gene Name: | DAN polymerase delta 1, catalytic subunit |
Gene Aliases: |
CDC2, CRCS10, MDPL, POLD |
Location: |
Chr.19:50384291-50418018 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLD1 | NM_001256849.1 | NP_001243778.1 | ||
| NM_001308632.1 | NP_001295561.1 | |||
| NM_002691.3 | NP_002682.2 | |||
| NR_046402.1 | ||||
| XM_005259008.4 | XP_005259065.1 | |||
| XM_011527038.1 | XP_011525340.1 | |||
| XM_017026881.1 | XP_016882370.1 | |||
| XM_017026882.1 | XP_016882371.1 | |||
| AB209560.1 | BAD92797.1 | |||
| BC008800.1 | AAH08800.1 | |||
| DQ234348.1 | ABB29977.1 | |||
| M80397.1 | AAA58439.1 | |||
| M81735.1 | AAA35768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map