Assay Details
Target Gene Details
Entrez Gene ID: | 493861 |
Gene Name: | EP300 interacting inhibitor of differentiation 3 |
Gene Aliases: |
NS4EB, NSE4B, NSMCE4B |
Location: |
Chr.12:104303732-104305205 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EID3 | NM_001008394.2 | 1 | 39 | NP_001008395.1 |
| AK098698.1 | 1 | 32 | BAC05385.1 | |
| BC027612.2 | AAH27612.1 | |||
| BI461575.1 | 1 | 44 |
Target Gene Details
Entrez Gene ID: | 7296 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
GRIM-12, TR, TR1, TRXR1, TXNR |
Location: |
Chr.12:104215779-104350307 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv52994 | Chr.12:104302508 - 104307999 on Build GRCh38 | Deletion |
|
| esv3630634 | Chr.12:104247347 - 104343900 on Build GRCh38 | Gain |
|
| esv3580448 | Chr.12:104283545 - 104307551 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map