Assay Details
Target Gene Details
Entrez Gene ID: | 55739 |
Gene Name: | NAD(P)HX dehydratase |
Gene Aliases: |
CARKD, LP3298 |
Location: |
Chr.13:110615460-110639996 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAXD | NM_001242881.1 | 10 | 1262 | NP_001229810.1 |
| NM_001242882.1 | 10 | 1259 | NP_001229811.1 | |
| NM_001242883.1 | 7 | 973 | NP_001229812.1 | |
| NM_018210.3 | 10 | 1398 | NP_060680.2 | |
| NR_040103.1 | 10 | 1254 | ||
| NR_040104.1 | 9 | 1170 | ||
| AK001631.1 | 10 | 1321 | BAA91797.1 | |
| AK024260.1 | 9 | 1039 | BAB14863.1 | |
| AK057767.1 | ||||
| AK058043.1 | 1 | 2139 | ||
| AK091867.1 | 1 | 1326 | ||
| AK093771.1 | 10 | 1180 | ||
| AK225129.1 | 10 | 1184 | ||
| AK296758.1 | 11 | 977 | ||
| AK298917.1 | 6 | 790 | ||
| AK302117.1 | 7 | 858 | ||
| BC041028.1 | 10 | 1193 | AAH41028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456120 | Chr.13:110627655 - 110657270 on Build GRCh38 | Loss |
|
| nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain |
|
| nsv563124 | Chr.13:110608920 - 110648596 on Build GRCh38 | Loss |
|
| nsv832720 | Chr.13:110447888 - 110643646 on Build GRCh38 | Loss |
|
| nsv563123 | Chr.13:110584913 - 110639697 on Build GRCh38 | Loss |
|
| nsv1042232 | Chr.13:110629776 - 110696128 on Build GRCh38 | Gain |
|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map