Assay Details
Target Gene Details
Entrez Gene ID: | 7047 |
Gene Name: | transglutaminase 4 |
Gene Aliases: |
TGP, hTGP |
Location: |
Chr.3:44874592-44914601 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM4 | NM_003241.3 | NP_003232.2 | ||
| XM_011534042.1 | XP_011532344.1 | |||
| AK125536.1 | ||||
| AK300523.1 | ||||
| AK312936.1 | ||||
| BC007003.1 | AAH07003.1 | |||
| DA870110.1 | ||||
| L34840.1 | AAB95430.1 | |||
| U31905.1 | AAC50516.1 | |||
| U79008.1 | AAB38324.1 | |||
| Y09559.1 | CAA70753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3792 | Chr.3:44836688 - 44881327 on Build GRCh38 | Deletion |
|
| nsv979826 | Chr.3:44872532 - 44874641 on Build GRCh38 | Duplication |
|
| nsv834675 | Chr.3:44805220 - 44966381 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map