Assay Details
Target Gene Details
Entrez Gene ID: | 23632 |
Gene Name: | carbonic anhydrase 14 |
Gene Aliases: |
CAXiV |
Location: |
Chr.1:150257255-150265078 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA14 | NM_012113.2 | 11 | 1471 | NP_036245.1 |
| XM_005245059.2 | 11 | 1397 | XP_005245116.1 | |
| XM_005245060.2 | 10 | 1315 | XP_005245117.1 | |
| XM_006711259.2 | 10 | 1312 | XP_006711322.1 | |
| XM_006711261.2 | 11 | 1840 | XP_006711324.1 | |
| XM_011509379.2 | 11 | 1917 | XP_011507681.1 | |
| XM_017000893.1 | 10 | 1755 | XP_016856382.1 | |
| XM_017000894.1 | 9 | 1175 | XP_016856383.1 | |
| XM_017000895.1 | 8 | 1090 | XP_016856384.1 | |
| XM_017000896.1 | 9 | 1615 | XP_016856385.1 | |
| AB025904.1 | 11 | 1394 | BAA85002.1 | |
| AK074765.1 | 11 | 1440 | BAC11191.1 | |
| AK290609.1 | 11 | 1310 | ||
| AY358689.1 | 11 | 1355 | AAQ89052.1 | |
| BC034412.1 | 11 | 1325 | AAH34412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009508 | Chr.1:150238763 - 150298405 on Build GRCh38 | Loss |
|
| nsv1011223 | Chr.1:150256810 - 150306949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map