Assay Details
Target Gene Details
Entrez Gene ID: | 64750 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 2 |
Gene Aliases: |
- |
Location: |
Chr.17:64542282-64662268 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMURF2 | NM_022739.3 | 19 | 3322 | NP_073576.1 |
| XM_005257585.3 | 18 | 2922 | XP_005257642.1 | |
| XM_011525128.1 | 19 | 2940 | XP_011523430.1 | |
| AK002019.1 | 1 | 265 | ||
| CR936721.1 | 20 | 3057 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066765 | Chr.17:64520760 - 65128185 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map