Assay Details
Target Gene Details
Entrez Gene ID: | 28976 |
Gene Name: | acyl-CoA dehydrogenase family member 9 |
Gene Aliases: |
NPD002 |
Location: |
Chr.3:128879490-128916075 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAD9 | NM_014049.4 | NP_054768.2 | ||
| NR_033426.1 | ||||
| AB209427.1 | BAD92664.1 | |||
| AF078854.1 | AAD44486.1 | |||
| AF327351.1 | AAL56011.1 | |||
| AK024012.1 | BAB14775.1 | |||
| AK027186.1 | ||||
| AK130311.1 | ||||
| BC007970.2 | AAH07970.1 | |||
| BC013354.2 | AAH13354.1 |
Target Gene Details
Entrez Gene ID: | 57501 |
Gene Name: | KIAA1257 |
Gene Aliases: |
- |
Location: |
Chr.3:128910186-129032431 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1257 | XM_011513050.2 | XP_011511352.2 | ||
| XM_017006939.1 | 16 | 5919 | XP_016862428.1 | |
| XM_017006940.1 | 16 | 5915 | XP_016862429.1 | |
| XM_017006941.1 | 16 | 5492 | XP_016862430.1 | |
| XM_017006942.1 | 16 | 5886 | XP_016862431.1 | |
| XM_017006943.1 | 16 | 5644 | XP_016862432.1 | |
| XM_017006944.1 | 15 | 5464 | XP_016862433.1 | |
| XM_017006945.1 | XP_016862434.1 | |||
| XM_017006946.1 | 14 | 5075 | XP_016862435.1 | |
| XM_017006947.1 | 13 | 4959 | XP_016862436.1 | |
| XM_017006948.1 | 15 | 5136 | XP_016862437.1 | |
| XM_017006949.1 | 14 | 5083 | XP_016862438.1 | |
| XM_017006950.1 | 14 | 10570 | XP_016862439.1 | |
| XM_017006951.1 | 14 | 10675 | XP_016862440.1 | |
| XM_017006952.1 | 13 | 4853 | XP_016862441.1 | |
| XM_017006953.1 | 13 | 4859 | XP_016862442.1 | |
| XM_017006954.1 | XP_016862443.1 |
More Information
Set Membership: |
|

Genomic Map