Assay Details
Target Gene Details
Entrez Gene ID: | 5479 |
Gene Name: | peptidylprolyl isomerase B |
Gene Aliases: |
CYP-S1, CYPB, HEL-S-39, OI9, SCYLP |
Location: |
Chr.15:64155815-64163155 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIB | NM_000942.4 | NP_000933.1 | ||
| AK291149.1 | ||||
| AK311074.1 | ||||
| BC001125.2 | AAH01125.1 | |||
| BC008848.2 | AAH08848.1 | |||
| BC020800.1 | AAH20800.1 | |||
| BC032138.2 | AAH32138.1 | |||
| BQ931949.1 | ||||
| CR456829.1 | CAG33110.1 | |||
| EU794651.1 | ||||
| M60457.1 | AAA35733.1 | |||
| M60857.1 | AAA52150.1 | |||
| M63573.1 | AAA36601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833036 | Chr.15:64072492 - 64215189 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map