Assay Details
Target Gene Details
Entrez Gene ID: | 9730 |
Gene Name: | DDB1 and CUL4 associated factor 1 |
Gene Aliases: |
VPRBP |
Location: |
Chr.3:51395867-51505620 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF1 | NM_001171904.1 | NP_001165375.1 | ||
| NM_014703.2 | NP_055518.1 | |||
| XM_005276753.4 | XP_005276810.1 | |||
| XM_005276755.4 | XP_005276812.1 | |||
| XM_011534273.2 | XP_011532575.1 | |||
| XM_011534274.2 | XP_011532576.1 | |||
| XM_011534275.2 | XP_011532577.1 | |||
| XM_011534276.2 | XP_011532578.1 | |||
| XM_011534277.2 | XP_011532579.1 | |||
| XM_017007546.1 | XP_016863035.1 | |||
| XM_017007547.1 | XP_016863036.1 | |||
| XM_017007548.1 | XP_016863037.1 | |||
| XM_017007549.1 | XP_016863038.1 | |||
| XM_017007550.1 | XP_016863039.1 | |||
| XM_017007551.1 | XP_016863040.1 | |||
| AB018343.1 | ||||
| AF061935.1 | AAG27134.1 | |||
| AK023239.1 | ||||
| AL080145.1 | CAB45738.1 | |||
| BC022792.1 | AAH22792.1 | |||
| BC110371.1 | AAI10372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590277 | Chr.3:51370620 - 51466575 on Build GRCh38 | Loss |
|
| nsv590276 | Chr.3:51358140 - 51431657 on Build GRCh38 | Loss |
|
| nsv3816 | Chr.3:51402359 - 51443460 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map