Assay Details
Target Gene Details
Entrez Gene ID: | 389791 |
Gene Name: | PTGES2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.9:128128529-128130634 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES2-AS1 | NR_024425.1 | 1 | 567 | |
| AK094537.1 | 1 | 554 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951769 | Chr.9:128101422 - 128129321 on Build GRCh38 | Deletion |
|
| nsv6723 | Chr.9:128106647 - 128141731 on Build GRCh38 | Deletion |
|
| nsv471324 | Chr.9:128039877 - 128130666 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145557555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map