Assay Details
Target Gene Details
Entrez Gene ID: | 8405 |
Gene Name: | speckle type BTB/POZ protein |
Gene Aliases: |
BTBD32, TEF2 |
Location: |
Chr.17:49598884-49678163 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOP | NM_001007226.1 | 12 | 1888 | NP_001007227.1 |
| NM_001007227.1 | 11 | 1766 | NP_001007228.1 | |
| NM_001007228.1 | 10 | 1714 | NP_001007229.1 | |
| NM_001007229.1 | 11 | 1765 | NP_001007230.1 | |
| NM_001007230.1 | 12 | 1817 | NP_001007231.1 | |
| NM_003563.3 | 11 | 1836 | NP_003554.1 | |
| XM_005257723.4 | 11 | 2060 | XP_005257780.1 | |
| XM_005257724.4 | 10 | 1990 | XP_005257781.1 | |
| XM_017025204.1 | 9 | 1503 | XP_016880693.1 | |
| AJ000644.1 | 10 | 1574 | CAA04199.1 | |
| AK123385.1 | 1 | 1546 | ||
| AK222589.1 | 10 | 1539 | BAD96309.1 | |
| AK293587.1 | 10 | 1520 | ||
| AK303268.1 | ||||
| BC001269.1 | 11 | 1720 | AAH01269.1 | |
| BC003385.1 | 10 | 1531 | AAH03385.1 | |
| BM693397.1 | 1 | 242 | ||
| W84567.1 | 1 | 193 |
More Information
Set Membership: |
|

Genomic Map