Assay Details
Target Gene Details
Entrez Gene ID: | 29980 |
Gene Name: | downstream neighbor of SON |
Gene Aliases: |
B17, C21orf60 |
Location: |
Chr.21:33577551-33588708 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DONSON | NM_017613.3 | 10 | 1737 | NP_060083.1 |
| AF232673.1 | 9 | 1358 | AAF72947.1 | |
| AF232674.1 | 8 | 1179 | AAF72948.1 | |
| AF232675.1 | 5 | 793 | AAF72949.1 | |
| AK001274.1 | ||||
| AK074964.1 | 10 | 1686 | BAC11320.1 | |
| BC048266.1 | 10 | 1687 | AAH48266.1 |
More Information
Set Membership: |
|

Genomic Map