Assay Details
Target Gene Details
Entrez Gene ID: | 102465464 |
Gene Name: | microRNA 6775 |
Gene Aliases: |
hsa-mir-6775 |
Location: |
Chr.16:87834592-87834660 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6775 | NR_106833.1 |
Target Gene Details
Entrez Gene ID: | 8140 |
Gene Name: | solute carrier family 7 member 5 |
Gene Aliases: |
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1 |
Location: |
Chr.16:87830022-87869499 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A5 | NM_003486.5 | NP_003477.4 | ||
| XM_006721286.3 | XP_006721349.2 | |||
| XM_017023735.1 | XP_016879224.1 | |||
| XM_017023736.1 | XP_016879225.1 | |||
| XM_017023737.1 | XP_016879226.1 | |||
| AB017908.1 | BAA75746.1 | |||
| AB018009.1 | BAA84648.1 | |||
| AB018542.1 | BAA33851.1 | |||
| AF077866.1 | AAC61479.1 | |||
| AF104032.1 | AAD20464.1 | |||
| BC039692.2 | AAH39692.1 | |||
| BC042600.1 | AAH42600.1 | |||
| M80244.1 | AAA35780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059786 | Chr.16:87811419 - 88019505 on Build GRCh38 | Gain |
|
| nsv833322 | Chr.16:87715029 - 87884870 on Build GRCh38 | Loss |
|
| nsv1160441 | Chr.16:87761248 - 87867260 on Build GRCh38 | Deletion |
|
| esv3892935 | Chr.16:87808912 - 88010269 on Build GRCh38 | Loss |
|
| dgv3062n100 | Chr.16:87725235 - 87842257 on Build GRCh38 | Loss |
|
| nsv573557 | Chr.16:87808535 - 87854711 on Build GRCh38 | Loss |
|
| nsv1062946 | Chr.16:87635801 - 88019505 on Build GRCh38 | Gain |
|
| dgv36n68 | Chr.16:87801736 - 87983550 on Build GRCh38 | Loss |
|
| nsv573536 | Chr.16:87705252 - 87914409 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map