Assay Details
Target Gene Details
Entrez Gene ID: | 2804 |
Gene Name: | golgin B1 |
Gene Aliases: |
GCP, GCP372, GOLIM1 |
Location: |
Chr.3:121663199-121749908 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGB1 | NM_001256486.1 | 22 | 9926 | NP_001243415.1 |
| NM_001256487.1 | 22 | 9939 | NP_001243416.1 | |
| NM_001256488.1 | 21 | 9816 | NP_001243417.1 | |
| NM_004487.4 | 22 | 9896 | NP_004478.3 | |
| XM_005247371.4 | 22 | 10386 | XP_005247428.1 | |
| XM_005247372.4 | 22 | 9962 | XP_005247429.1 | |
| XM_005247373.1 | 21 | 9778 | XP_005247430.1 | |
| XM_006713587.1 | 22 | 9886 | XP_006713650.1 | |
| XM_006713588.1 | 22 | 9886 | XP_006713651.1 | |
| XM_006713589.1 | 22 | 9883 | XP_006713652.1 | |
| XM_006713590.1 | 21 | 9763 | XP_006713653.1 | |
| XM_006713591.1 | 20 | 9661 | XP_006713654.1 | |
| XM_011512699.2 | 22 | 9853 | XP_011511001.1 | |
| XM_017006189.1 | 22 | 10371 | XP_016861678.1 | |
| XM_017006190.1 | 22 | 10538 | XP_016861679.1 | |
| XM_017006191.1 | 21 | 10263 | XP_016861680.1 | |
| XM_017006192.1 | 22 | 9947 | XP_016861681.1 | |
| XM_017006193.1 | 22 | 9944 | XP_016861682.1 | |
| XM_017006194.1 | 21 | 9839 | XP_016861683.1 | |
| XM_017006195.1 | 16 | 9808 | XP_016861684.1 | |
| AB371588.1 | 22 | 9899 | ||
| AB593126.1 | 21 | 9790 | ||
| AK094551.1 | 1 | 2736 | ||
| BF061034.1 | 1 | 400 | ||
| BX537520.1 | 1 | 2872 | ||
| D25542.1 | 22 | 9894 | BAA05025.1 | |
| X75304.1 | 22 | 9884 | CAA53052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain |
|
| esv3575599 | Chr.3:121571453 - 121671424 on Build GRCh38 | Gain |
|
| nsv829703 | Chr.3:121664443 - 121815646 on Build GRCh38 | Loss |
|
| nsv999335 | Chr.3:121381436 - 121689919 on Build GRCh38 | Gain |
|
| esv3575598 | Chr.3:121406926 - 121715587 on Build GRCh38 | Gain |
|
| nsv998740 | Chr.3:121570652 - 121671424 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map