Assay Details
Target Gene Details
Entrez Gene ID: | 60625 |
Gene Name: | DEAH-box helicase 35 |
Gene Aliases: |
C20orf15, DDX35, KAIA0875 |
Location: |
Chr.20:38962338-39039723 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX35 | NM_001190809.1 | NP_001177738.1 | ||
| NM_021931.3 | NP_068750.2 | |||
| NR_033905.1 | ||||
| XM_011528972.1 | XP_011527274.1 | |||
| XM_011528974.1 | XP_011527276.1 | |||
| AK026412.1 | BAB15476.1 | |||
| AK296506.1 | ||||
| AK296793.1 | ||||
| AK303396.1 | ||||
| AK307863.1 | ||||
| BC132669.1 | ||||
| BG258573.1 | ||||
| DA725122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645798 | Chr.20:38949859 - 39066865 on Build GRCh38 | Loss |
|
| esv3645793 | Chr.20:38918552 - 38972807 on Build GRCh38 | Gain |
|
| esv3645796 | Chr.20:38949708 - 38974834 on Build GRCh38 | Loss |
|
| esv3645794 | Chr.20:38934425 - 39003285 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map