Assay Details
Target Gene Details
Entrez Gene ID: | 83401 |
Gene Name: | ELOVL fatty acid elongase 3 |
Gene Aliases: |
CIG-30, CIG30 |
Location: |
Chr.10:102224767-102229589 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL3 | NM_152310.2 | 1 | 165 | NP_689523.1 |
| XM_011540245.2 | XP_011538547.1 | |||
| BC034344.1 | 1 | 57 | AAH34344.1 | |
| HY050356.1 | 1 | 166 |
More Information
Set Membership: |
|

Genomic Map