Assay Details
Target Gene Details
Entrez Gene ID: | 84947 |
Gene Name: | serine active site containing 1 |
Gene Aliases: |
- |
Location: |
Chr.6:158109504-158168280 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERAC1 | NM_032861.3 | 17 | 3905 | NP_116250.3 |
| XM_006715586.2 | 16 | 3834 | XP_006715649.1 | |
| XM_011536196.2 | 16 | 3928 | XP_011534498.1 | |
| XM_011536198.2 | 15 | 3830 | XP_011534500.1 | |
| AA128978.1 | 1 | 65 | ||
| AK023656.1 | 1 | 1941 | ||
| BC001705.1 | 17 | 3862 | AAH01705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890972 | Chr.6:158082895 - 158303958 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113233603] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map