Assay Details
Target Gene Details
Entrez Gene ID: | 56114 |
Gene Name: | protocadherin gamma subfamily A, 1 |
Gene Aliases: |
PCDH-GAMMA-A1 |
Location: |
Chr.5:141330685-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA1 | NM_018912.2 | NP_061735.1 | ||
| AF152318.1 | AAD43712.1 |
Target Gene Details
Entrez Gene ID: | 56113 |
Gene Name: | protocadherin gamma subfamily A, 2 |
Gene Aliases: |
PCDH-GAMMA-A2 |
Location: |
Chr.5:141338760-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA2 | NM_018915.3 | NP_061738.1 | ||
| AF152322.1 | AAD43716.1 |
Target Gene Details
Entrez Gene ID: | 56112 |
Gene Name: | protocadherin gamma subfamily A, 3 |
Gene Aliases: |
PCDH-GAMMA-A3 |
Location: |
Chr.5:141344034-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA3 | NM_018916.3 | NP_061739.2 | ||
| AB208927.1 | BAD92164.1 | |||
| AF152323.1 | AAD43717.1 |
Target Gene Details
Entrez Gene ID: | 56111 |
Gene Name: | protocadherin gamma subfamily A, 4 |
Gene Aliases: |
PCDH-GAMMA-A4 |
Location: |
Chr.5:141355025-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA4 | NM_018917.3 | NP_061740.2 | ||
| AF152324.1 | AAD43718.1 |
Target Gene Details
Entrez Gene ID: | 56110 |
Gene Name: | protocadherin gamma subfamily A, 5 |
Gene Aliases: |
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5 |
Location: |
Chr.5:141364331-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA5 | NM_018918.2 | NP_061741.1 | ||
| AF152325.1 | AAD43719.1 |
Target Gene Details
Entrez Gene ID: | 56109 |
Gene Name: | protocadherin gamma subfamily A, 6 |
Gene Aliases: |
PCDH-GAMMA-A6 |
Location: |
Chr.5:141374084-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA6 | NM_018919.2 | NP_061742.1 | ||
| AB209178.1 | BAD92415.1 | |||
| AF152326.1 | AAD43720.1 | |||
| BC146651.1 |
Target Gene Details
Entrez Gene ID: | 56108 |
Gene Name: | protocadherin gamma subfamily A, 7 |
Gene Aliases: |
PCDH-GAMMA-A7 |
Location: |
Chr.5:141382739-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA7 | NM_018920.3 | NP_061743.1 | ||
| AB208958.1 | BAD92195.1 | |||
| AF152327.1 | AAD43721.1 |
Target Gene Details
Entrez Gene ID: | 56104 |
Gene Name: | protocadherin gamma subfamily B, 1 |
Gene Aliases: |
PCDH-GAMMA-B1 |
Location: |
Chr.5:141350261-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB1 | NM_018922.2 | NP_061745.1 | ||
| AF152330.1 | AAD43724.1 |
Target Gene Details
Entrez Gene ID: | 56103 |
Gene Name: | protocadherin gamma subfamily B, 2 |
Gene Aliases: |
PCDH-GAMMA-B2 |
Location: |
Chr.5:141360136-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB2 | NM_018923.2 | NP_061746.1 | ||
| AF152331.1 | AAD43725.1 |
Target Gene Details
Entrez Gene ID: | 56102 |
Gene Name: | protocadherin gamma subfamily B, 3 |
Gene Aliases: |
PCDH-GAMMA-B3 |
Location: |
Chr.5:141370264-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB3 | NM_018924.4 | NP_061747.2 | ||
| AF152332.1 | AAD43726.1 | |||
| BC150172.1 |
Target Gene Details
Entrez Gene ID: | 8641 |
Gene Name: | protocadherin gamma subfamily B, 4 |
Gene Aliases: |
CDH20, FIB2, PCDH-GAMMA-B4 |
Location: |
Chr.5:141387885-141512979 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB4 | NM_003736.2 | 1 | 859 | NP_003727.1 |
| NM_032098.1 | 1 | 859 | NP_115269.1 | |
| AF152333.1 | 1 | 859 | AAD43727.1 | |
| AF152520.1 | 1 | 859 | AAD43780.1 | |
| BC112088.1 | 1 | 906 | AAI12089.1 | |
| BC113607.1 | 1 | 908 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv599922 | Chr.5:141341879 - 141416532 on Build GRCh38 | Loss |
|
| nsv823280 | Chr.5:141388318 - 141397872 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map