Assay Details
Target Gene Details
Entrez Gene ID: | 84299 |
Gene Name: | migration and invasion enhancer 1 |
Gene Aliases: |
C17orf37, C35, ORB3, RDX12, XTP4 |
Location: |
Chr.17:39728500-39730563 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIEN1 | NM_032339.4 | NP_115715.3 | ||
| XM_005257736.3 | XP_005257793.1 | |||
| AF490253.1 | AAO85461.1 | |||
| AJ308025.1 | ||||
| AJ308026.1 | ||||
| AL554581.3 | ||||
| AY508814.1 | AAR92035.1 | |||
| BC006006.1 | AAH06006.1 | |||
| BX646129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056234 | Chr.17:39706352 - 39753735 on Build GRCh38 | Gain |
|
| nsv2048 | Chr.17:39718682 - 39742588 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map