Assay Details
Target Gene Details
Entrez Gene ID: | 4613 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog |
Gene Aliases: |
MODED, N-myc, NMYC, ODED, bHLHe37 |
Location: |
Chr.2:15940438-15947007 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCN | NM_001293228.1 | 3 | 1932 | NP_001280157.1 |
| NM_001293231.1 | 2 | 715 | NP_001280160.1 | |
| NM_001293233.1 | 3 | 1622 | NP_001280162.1 | |
| NM_005378.5 | 3 | 1622 | NP_005369.2 | |
| XM_017004168.1 | 2 | 1453 | XP_016859657.1 | |
| AK298225.1 | 4 | 915 | ||
| AK299495.1 | 4 | 1250 | ||
| BC002712.2 | 2 | 1340 | AAH02712.1 | |
| BT007384.1 | 2 | 1199 | AAP36048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1102e212 | Chr.2:15937643 - 15955551 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map