Assay Details
Target Gene Details
Entrez Gene ID: | 131 |
Gene Name: | alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide |
Gene Aliases: |
ADH4 |
Location: |
Chr.4:99412261-99435510 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH7 | NM_000673.4 | 7 | 1193 | NP_000664.2 |
| NM_001166504.1 | 7 | 1006 | NP_001159976.1 | |
| AK223472.1 | 7 | 987 | ||
| AK301657.1 | 7 | 969 | ||
| AK301696.1 | 7 | 1006 | ||
| AK310811.1 | ||||
| AK312854.1 | 7 | 1116 | ||
| BC131512.1 | 7 | 988 | ||
| CD014081.1 | 8 | 1069 | ||
| L33179.1 | 7 | 916 | AAA59211.1 | |
| S77168.1 | AAB34478.1 | |||
| U07821.1 | 7 | 1000 | AAA19002.1 | |
| U09623.1 | 7 | 934 | AAA82165.1 | |
| X76342.1 | 7 | 976 | CAA53960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517344 | Chr.4:99401288 - 99442412 on Build GRCh38 | Gain |
|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| esv3584519 | Chr.4:99402713 - 99556305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map