Assay Details
Target Gene Details
Entrez Gene ID: | 4486 |
Gene Name: | macrophage stimulating 1 receptor |
Gene Aliases: |
CD136, CDw136, PTK8, RON |
Location: |
Chr.3:49886471-49903637 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MST1R | NM_001244937.2 | 19 | 4080 | NP_001231866.1 |
| NM_001318913.1 | 18 | 3909 | NP_001305842.1 | |
| NM_002447.3 | 20 | 4227 | NP_002438.2 | |
| NR_134919.1 | 20 | 4163 | ||
| XM_005265170.4 | 20 | 4225 | XP_005265227.2 | |
| XM_011533739.2 | 20 | 4105 | XP_011532041.1 | |
| XM_011533740.2 | 19 | 4078 | XP_011532042.1 | |
| XM_011533741.2 | 19 | 4018 | XP_011532043.1 | |
| XM_011533742.2 | 19 | 4088 | XP_011532044.1 | |
| XM_011533743.2 | 18 | 3922 | XP_011532045.1 | |
| XM_011533744.2 | 18 | 3907 | XP_011532046.1 | |
| AK303234.1 | 10 | 1567 | ||
| CA503122.1 | 1 | 323 | ||
| HQ908652.1 | ||||
| JN689381.1 | ||||
| JN689382.1 | ||||
| JN983806.1 | 18 | 3909 | ||
| X70040.1 | 20 | 4227 | CAA49634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834688 | Chr.3:49773682 - 49981866 on Build GRCh38 | Loss |
|
| dgv890e214 | Chr.3:49882736 - 49888371 on Build GRCh38 | Loss |
|
| esv3596133 | Chr.3:49882993 - 49887598 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map